RIE156325098 Rieke Metals, Inc.

2,3-dimethyl-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842366
  • Cas Number : 898790-13-1
  • Molecular Weight : 291.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-26
Availability details for
Quantity
  • Availability
  • Add to Cart