RIE156325101 Rieke Metals, Inc.

2,6-dimethyl-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842369
  • Cas Number : 898790-22-2
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl](2,6-dimethylphenyl)methanone
Inchi Code 1S/C20H21NO/c1-15-7-5-8-16(2)19(15)20(22)18-10-6-9-17(13-18)14-21-11-3-4-12-21/h3-10,13H,11-12,14H2,1-2H3
Inchi Key CWUVQJCZECFPHH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0900-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-29
Availability details for
Quantity
  • Availability
  • Add to Cart