RIE156325104 Rieke Metals, Inc.

4-bromo-3-fluoro-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842372
  • Cas Number : 898749-10-5
  • Molecular Weight : 360.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-32
Availability details for
Quantity
  • Availability
  • Add to Cart