RIE156325110 Rieke Metals, Inc.

3-(3-pyrrolinomethyl)-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842378
  • Cas Number : 898749-29-6
  • Molecular Weight : 331.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-44
Availability details for
Quantity
  • Availability
  • Add to Cart