RIE156325117 Rieke Metals, Inc.

2,4-dichloro-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842385
  • Cas Number : 898749-50-3
  • Molecular Weight : 332.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-dichlorophenyl)[3-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]methanone
Inchi Code 1S/C18H15Cl2NO/c19-15-6-7-16(17(20)11-15)18(22)14-5-3-4-13(10-14)12-21-8-1-2-9-21/h1-7,10-11H,8-9,12H2
Inchi Key RSBBFRCXJBLGDW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0900-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-54
Availability details for
Quantity
  • Availability
  • Add to Cart