RIE156325121 Rieke Metals, Inc.

2,4-difluoro-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842389
  • Cas Number : 898749-58-1
  • Molecular Weight : 299.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluorophenyl)[3-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]methanone
Inchi Code 1S/C18H15F2NO/c19-15-6-7-16(17(20)11-15)18(22)14-5-3-4-13(10-14)12-21-8-1-2-9-21/h1-7,10-11H,8-9,12H2
Inchi Key NHHKKKLKNJVPGJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0900-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-58
Availability details for
Quantity
  • Availability
  • Add to Cart