RIE156325124 Rieke Metals, Inc.

3'-(3-pyrrolinomethyl)-3,4,5-trifluorobenzophenone

  • MDL number MFCD03842392
  • Cas Number : 898749-64-9
  • Molecular Weight : 317.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-61
Availability details for
Quantity
  • Availability
  • Add to Cart