RIE156325125 Rieke Metals, Inc.

Cyclopropyl 3-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03842393
  • Cas Number : 898749-66-1
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0901-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0901-00
Availability details for
Quantity
  • Availability
  • Add to Cart