RIE156325126 Rieke Metals, Inc.

Cyclobutyl 3-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03842394
  • Cas Number : 898749-69-4
  • Molecular Weight : 241.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0902-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0902-00
Availability details for
Quantity
  • Availability
  • Add to Cart