RIE156325127 Rieke Metals, Inc.

Cyclopentyl 3-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03842395
  • Cas Number : 898749-72-9
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0903-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0903-00
Availability details for
Quantity
  • Availability
  • Add to Cart