RIE156325128 Rieke Metals, Inc.

Cyclohexyl 3-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03842396
  • Cas Number : 898749-74-1
  • Molecular Weight : 269.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0904-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0904-00
Availability details for
Quantity
  • Availability
  • Add to Cart