RIE156325131 Rieke Metals, Inc.

Ethyl 6-oxo-6-[3-(3-pyrrolinomethyl)phenyl]hexanoate

  • MDL number MFCD03842399
  • Cas Number : 898749-80-9
  • Molecular Weight : 315.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0907-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0907-00
Availability details for
Quantity
  • Availability
  • Add to Cart