RIE156325135 Rieke Metals, Inc.

2-Methyl-2'-morpholinomethyl benzophenone

  • MDL number MFCD03842403
  • Cas Number : 898749-87-6
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-02
Availability details for
Quantity
  • Availability
  • Add to Cart