RIE156325136 Rieke Metals, Inc.

3'-Methyl-2-morpholinomethyl benzophenone

  • MDL number MFCD03842404
  • Cas Number : 898749-90-1
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-03
Availability details for
Quantity
  • Availability
  • Add to Cart