RIE156325146 Rieke Metals, Inc.

4'-Carboethoxy-2-morpholinomethyl benzophenone

  • MDL number MFCD03842414
  • Cas Number : 898750-20-4
  • Molecular Weight : 353.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-13
Availability details for
Quantity
  • Availability
  • Add to Cart