RIE156325153 Rieke Metals, Inc.

3'-Fluoro-2-morpholinomethyl benzophenone

  • MDL number MFCD03842421
  • Cas Number : 898750-41-9
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-22
Availability details for
Quantity
  • Availability
  • Add to Cart