RIE156325160 Rieke Metals, Inc.

3',5'-Dimethyl-2-morpholinomethyl benzophenone

  • MDL number MFCD03842428
  • Cas Number : 898750-59-9
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-31
Availability details for
Quantity
  • Availability
  • Add to Cart