RIE156325163 Rieke Metals, Inc.

3'-Chloro-4'-fluoro-2-morpholinomethyl benzophenone

  • MDL number MFCD03842431
  • Cas Number : 898750-68-0
  • Molecular Weight : 333.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-34
Availability details for
Quantity
  • Availability
  • Add to Cart