RIE156325164 Rieke Metals, Inc.

2-Chloro-2'-morpholinomethyl benzophenone

  • MDL number MFCD03842432
  • Cas Number : 898750-71-5
  • Molecular Weight : 315.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-36
Availability details for
Quantity
  • Availability
  • Add to Cart