RIE156325166 Rieke Metals, Inc.

2-Morpholinomethyl-2'-trifluoromethylbenzophenone

  • MDL number MFCD03842434
  • Cas Number : 898750-77-1
  • Molecular Weight : 349.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-43
Availability details for
Quantity
  • Availability
  • Add to Cart