RIE156325168 Rieke Metals, Inc.

2-Morpholinomethyl-4'-trifluoromethylbenzophenone

  • MDL number MFCD03842436
  • Cas Number : 898750-83-9
  • Molecular Weight : 349.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-45
Availability details for
Quantity
  • Availability
  • Add to Cart