RIE156325173 Rieke Metals, Inc.

2,3-Dichloro-2'-morpholinomethyl benzophenone

  • MDL number MFCD03842441
  • Cas Number : 898750-98-6
  • Molecular Weight : 350.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dichlorophenyl)[2-(4-morpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17Cl2NO2/c19-16-7-3-6-15(17(16)20)18(22)14-5-2-1-4-13(14)12-21-8-10-23-11-9-21/h1-7H,8-12H2
Inchi Key KZEUDOSQAVCHPN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0100-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-53
Availability details for
Quantity
  • Availability
  • Add to Cart