RIE156325174 Rieke Metals, Inc.

2,4-Dichloro-2'-morpholinomethyl benzophenone

  • MDL number MFCD03842442
  • Cas Number : 898751-01-4
  • Molecular Weight : 350.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-54
Availability details for
Quantity
  • Availability
  • Add to Cart