RIE156325176 Rieke Metals, Inc.

3',4'-Dichloro-2-morpholinomethyl benzophenone

  • MDL number MFCD03842444
  • Cas Number : 24088-69-5
  • Molecular Weight : 350.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-56
Availability details for
Quantity
  • Availability
  • Add to Cart