RIE156325179 Rieke Metals, Inc.

3',4'-Difluoro-2-morpholinomethyl benzophenone

  • MDL number MFCD03842447
  • Cas Number : 898751-17-2
  • Molecular Weight : 317.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)[2-(4-morpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17F2NO2/c19-16-6-5-13(11-17(16)20)18(22)15-4-2-1-3-14(15)12-21-7-9-23-10-8-21/h1-6,11H,7-10,12H2
Inchi Key RYQAPUHUKQCXLZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0100-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-59
Availability details for
Quantity
  • Availability
  • Add to Cart