RIE156325180 Rieke Metals, Inc.

3',5'-Difluoro-2-morpholinomethyl benzophenone

  • MDL number MFCD03842448
  • Cas Number : 898751-21-8
  • Molecular Weight : 317.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0100-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-60
Availability details for
Quantity
  • Availability
  • Add to Cart