RIE156325181 Rieke Metals, Inc.

2-Morpholinomethyl-3',4',5'-trifluorobenzophenone

  • MDL number MFCD03842449
  • Cas Number : 898751-25-2
  • Molecular Weight : 335.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(4-morpholinylmethyl)phenyl](3,4,5-trifluorophenyl)methanone
Inchi Code 1S/C18H16F3NO2/c19-15-9-13(10-16(20)17(15)21)18(23)14-4-2-1-3-12(14)11-22-5-7-24-8-6-22/h1-4,9-10H,5-8,11H2
Inchi Key VTQBEOFFPOIRHP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0100-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0100-61
Availability details for
Quantity
  • Availability
  • Add to Cart