RIE156325183 Rieke Metals, Inc.

Cyclobutyl 2-(morpholinomethyl)phenyl ketone

  • MDL number MFCD03842451
  • Cas Number : 898751-33-2
  • Molecular Weight : 259.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0102-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0102-00
Availability details for
Quantity
  • Availability
  • Add to Cart