RIE156325186 Rieke Metals, Inc.

Ethyl 5-[2-(morpholinomethyl)phenyl]-5-oxovalerate

  • MDL number MFCD03842455
  • Cas Number : 898751-49-0
  • Molecular Weight : 319.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0106-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0106-00
Availability details for
Quantity
  • Availability
  • Add to Cart