RIE156325191 Rieke Metals, Inc.

2-Methyl-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842460
  • Cas Number : 898751-65-0
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methylphenyl)[2-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO/c1-16-9-3-5-11-18(16)20(22)19-12-6-4-10-17(19)15-21-13-7-2-8-14-21/h3-6,9-12H,2,7-8,13-15H2,1H3
Inchi Key JIIAAIKHGBBOID-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-02
Availability details for
Quantity
  • Availability
  • Add to Cart