RIE156325193 Rieke Metals, Inc.

4'-Methyl-2-piperidinomethyl benzophenone

  • MDL number MFCD03842462
  • Cas Number : 898751-71-8
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-04
Availability details for
Quantity
  • Availability
  • Add to Cart