RIE156325197 Rieke Metals, Inc.

2-Cyano-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842466
  • Cas Number : 898751-83-2
  • Molecular Weight : 304.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-08
Availability details for
Quantity
  • Availability
  • Add to Cart