RIE156325200 Rieke Metals, Inc.

2-Carboethoxy-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842469
  • Cas Number : 898751-92-3
  • Molecular Weight : 351.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[2-(1-piperidinylmethyl)benzoyl]benzoate
Inchi Code 1S/C22H25NO3/c1-2-26-22(25)20-13-7-6-12-19(20)21(24)18-11-5-4-10-17(18)16-23-14-8-3-9-15-23/h4-7,10-13H,2-3,8-9,14-16H2,1H3
Inchi Key KONINNGYORTZQB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-11
Availability details for
Quantity
  • Availability
  • Add to Cart