RIE156325205 Rieke Metals, Inc.

3'-Bromo-2-piperidinomethyl benzophenone

  • MDL number MFCD03842474
  • Cas Number : 898773-03-0
  • Molecular Weight : 358.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)[2-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H20BrNO/c20-17-9-6-8-15(13-17)19(22)18-10-3-2-7-16(18)14-21-11-4-1-5-12-21/h2-3,6-10,13H,1,4-5,11-12,14H2
Inchi Key PPYBMWUCGNYYDW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-18
Availability details for
Quantity
  • Availability
  • Add to Cart