RIE156325206 Rieke Metals, Inc.

4'-Bromo-2-piperidinomethyl benzophenone

  • MDL number MFCD03842475
  • Cas Number : 898773-06-3
  • Molecular Weight : 358.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-19
Availability details for
Quantity
  • Availability
  • Add to Cart