RIE156325209 Rieke Metals, Inc.

3'-Fluoro-2-piperidinomethyl benzophenone

  • MDL number MFCD03842478
  • Cas Number : 898773-15-4
  • Molecular Weight : 297.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluorophenyl)[2-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H20FNO/c20-17-9-6-8-15(13-17)19(22)18-10-3-2-7-16(18)14-21-11-4-1-5-12-21/h2-3,6-10,13H,1,4-5,11-12,14H2
Inchi Key LIWBMXUBLWDKTJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-22
Availability details for
Quantity
  • Availability
  • Add to Cart