RIE156325211 Rieke Metals, Inc.

2,3-Dimethyl-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842480
  • Cas Number : 898773-21-2
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dimethylphenyl)[2-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C21H25NO/c1-16-9-8-12-19(17(16)2)21(23)20-11-5-4-10-18(20)15-22-13-6-3-7-14-22/h4-5,8-12H,3,6-7,13-15H2,1-2H3
Inchi Key APIQFZOYPAYLNR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-26
Availability details for
Quantity
  • Availability
  • Add to Cart