RIE156325215 Rieke Metals, Inc.

3',4'-Dimethyl-2-piperidinomethyl benzophenone

  • MDL number MFCD03842484
  • Cas Number : 898773-33-6
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-30
Availability details for
Quantity
  • Availability
  • Add to Cart