RIE156325218 Rieke Metals, Inc.

4'-Chloro-3'-fluoro-2-piperidinomethyl benzophenone

  • MDL number MFCD03842487
  • Cas Number : 898773-42-7
  • Molecular Weight : 331.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-33
Availability details for
Quantity
  • Availability
  • Add to Cart