RIE156325223 Rieke Metals, Inc.

2-Piperidinomethyl-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842492
  • Cas Number : 898773-53-0
  • Molecular Weight : 347.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-44
Availability details for
Quantity
  • Availability
  • Add to Cart