RIE156325224 Rieke Metals, Inc.

2-Piperidinomethyl-4'-trifluoromethylbenzophenone

  • MDL number MFCD03842493
  • Cas Number : 898773-55-2
  • Molecular Weight : 347.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(1-piperidinylmethyl)phenyl][4-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C20H20F3NO/c21-20(22,23)17-10-8-15(9-11-17)19(25)18-7-3-2-6-16(18)14-24-12-4-1-5-13-24/h2-3,6-11H,1,4-5,12-14H2
Inchi Key AQMRCFJQMKARNI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-45
Availability details for
Quantity
  • Availability
  • Add to Cart