RIE156325226 Rieke Metals, Inc.

2-Chloro-4-fluoro-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842495
  • Cas Number : 898773-59-6
  • Molecular Weight : 331.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-4-fluorophenyl)[2-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H19ClFNO/c20-18-12-15(21)8-9-17(18)19(23)16-7-3-2-6-14(16)13-22-10-4-1-5-11-22/h2-3,6-9,12H,1,4-5,10-11,13H2
Inchi Key GMUXAHGCQFFDFP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-50
Availability details for
Quantity
  • Availability
  • Add to Cart