RIE156325230 Rieke Metals, Inc.

2,4-dichloro-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842499
  • Cas Number : 898773-67-6
  • Molecular Weight : 348.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-54
Availability details for
Quantity
  • Availability
  • Add to Cart