RIE156325232 Rieke Metals, Inc.

3,4-dichloro-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842501
  • Cas Number : 898773-71-2
  • Molecular Weight : 348.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dichlorophenyl)[2-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H19Cl2NO/c20-17-9-8-14(12-18(17)21)19(23)16-7-3-2-6-15(16)13-22-10-4-1-5-11-22/h2-3,6-9,12H,1,4-5,10-11,13H2
Inchi Key TVPJJIXKYYPGIN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0200-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-56
Availability details for
Quantity
  • Availability
  • Add to Cart