RIE156325233 Rieke Metals, Inc.

3,5-dichloro-2'-piperidinomethyl benzophenone

  • MDL number MFCD03842502
  • Cas Number : 898773-73-4
  • Molecular Weight : 348.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0200-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0200-57
Availability details for
Quantity
  • Availability
  • Add to Cart