RIE156325239 Rieke Metals, Inc.

cyclobutyl 2-(piperidinomethyl)phenyl ketone

  • MDL number MFCD03842508
  • Cas Number : 898773-85-8
  • Molecular Weight : 257.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0202-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0202-00
Availability details for
Quantity
  • Availability
  • Add to Cart