RIE156325245 Rieke Metals, Inc.

ethyl 7-oxo-7-[2-(piperidinomethyl)phenyl]heptanoate

  • MDL number MFCD03842514
  • Cas Number : 898773-97-2
  • Molecular Weight : 345.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0208-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0208-00
Availability details for
Quantity
  • Availability
  • Add to Cart