RIE156325247 Rieke Metals, Inc.

2-(Pyrrolidinomethyl)benzophenone

  • MDL number MFCD03842516
  • Cas Number : 742000-78-8
  • Molecular Weight : 265.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-01
Availability details for
Quantity
  • Availability
  • Add to Cart