RIE156325251 Rieke Metals, Inc.

2-methoxy-2'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842520
  • Cas Number : 898774-11-3
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methoxyphenyl)[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H21NO2/c1-22-18-11-5-4-10-17(18)19(21)16-9-3-2-8-15(16)14-20-12-6-7-13-20/h2-5,8-11H,6-7,12-14H2,1H3
Inchi Key UPEUBFRAALBIIO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0300-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-05
Availability details for
Quantity
  • Availability
  • Add to Cart