RIE156325260 Rieke Metals, Inc.

2-pyrrolidinomethyl-2'-thiomethylbenzophenone

  • MDL number MFCD03842529
  • Cas Number : 898774-38-4
  • Molecular Weight : 311.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-14
Availability details for
Quantity
  • Availability
  • Add to Cart